Assay Details
Target Gene Details
Entrez Gene ID: | 68970 |
Gene Name: | DDB1 and CUL4 associated factor 12 |
Gene Aliases: |
1500001L20Rik, 5830424K06Rik, AA420338, AI851081, Wdr40a |
Location: |
Chr.4:41291300-41316726 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcaf12 | NM_026893.3 | NP_081169.2 | ||
AK005099.1 | BAB23817.2 | |||
AK030173.1 | BAC26822.1 | |||
AK031089.1 | BAC27247.1 | |||
AK078036.1 | ||||
AK087517.1 | BAC39908.1 | |||
AK090028.1 | BAC41053.1 | |||
AK143635.1 | ||||
AK173289.1 | BAD32567.1 | |||
BC023852.1 | AAH23852.1 |
