Assay Details
Target Gene Details
Entrez Gene ID: | 70832 |
Gene Name: | RIKEN cDNA 4921504A21 gene |
Gene Aliases: |
- |
Location: |
Chr.5:19202368-19226555 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4921504A21Rik | NR_102341.1 | |||
NR_102342.1 | ||||
AK014806.1 | ||||
AK046351.1 |
