Assay Details
Target Gene Details
Entrez Gene ID: | 21894 |
Gene Name: | talin 1 |
Gene Aliases: |
Tln |
Location: |
Chr.4:43531513-43562583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 34 - Exon 35 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tln1 | NM_011602.5 | NP_035732.2 | ||
AK122420.2 | BAC65702.4 | |||
AK147228.1 | BAE27781.1 | |||
AK170630.1 | BAE41923.1 | |||
AK171322.1 | BAE42391.1 | |||
BC121787.1 | ||||
BC150810.1 | ||||
X56123.1 | CAA39588.1 |
