Assay Details
Target Gene Details
Entrez Gene ID: | 21894 |
Gene Name: | talin 1 |
Gene Aliases: |
Tln |
Location: |
Chr.4:43531513-43562583 on Build GRCm38 |
Assay Gene Location: | Within Exon 37 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tln1 | NM_011602.5 | 37 | 5309 | NP_035732.2 |
AK122420.2 | 37 | 4935 | BAC65702.4 | |
AK147228.1 | 37 | 4880 | BAE27781.1 | |
AK170630.1 | 3 | 352 | BAE41923.1 | |
AK171322.1 | 3 | 352 | BAE42391.1 | |
BC018557.1 | AAH18557.1 | |||
BC121787.1 | 7 | 821 | ||
BC150810.1 | 37 | 4980 | ||
X56123.1 | 37 | 4981 | CAA39588.1 |
