Assay Details
Target Gene Details
Entrez Gene ID: | 85030 |
Gene Name: | tumor necrosis factor receptor superfamily, member 25 |
Gene Aliases: |
APO-3, DDR3, DR3, LARD, TR3, TRAMP, Tnfrsf12, WSL-1, WSL-LR, Wsl |
Location: |
Chr.4:152115528-152120119 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnfrsf25 | NM_001291010.1 | NP_001277939.1 | ||
NM_033042.4 | NP_149031.2 | |||
AF329969.1 | AAK11256.1 | |||
AK040632.1 | ||||
BC017526.1 | AAH17526.1 |
