Assay Details
Target Gene Details
Entrez Gene ID: | 69863 |
Gene Name: | tetratricopeptide repeat domain 39B |
Gene Aliases: |
1810054D07Rik, 9130422G05Rik, AI450603, AV103077, AW215585 |
Location: |
Chr.4:83220301-83324270 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc39b | NM_027238.2 | 4 | 313 | NP_081514.1 |
| AK007856.1 | 4 | 314 | BAB25308.1 | |
| AK037208.1 | 4 | 314 | BAC29754.1 | |
| AK049108.1 | 4 | 364 | BAC33545.1 | |
| BC151021.1 | 4 | 275 |

Genomic Map