Assay Details
Target Gene Details
Entrez Gene ID: | 67830 |
Gene Name: | RER1 retention in endoplasmic reticulum 1 homolog (S. cerevisiae) |
Gene Aliases: |
1110060F11Rik, 5830454N22Rik, AU043380 |
Location: |
Chr.4:155074109-155086382 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rer1 | NM_026395.1 | NP_080671.1 | ||
AK002550.1 | BAB22181.1 | |||
AK003682.1 | BAB22935.1 | |||
AK013262.1 | BAB28755.1 | |||
AK018013.1 | ||||
AK078396.1 | BAC37253.1 | |||
AK168192.1 | BAE40152.1 | |||
BC029189.1 | AAH29189.1 |
