Assay Details
Target Gene Details
Entrez Gene ID: | 231510 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 9 |
Gene Aliases: |
1-AGPAT, 1-AGPAT 9, 4933407I02Rik, 4933408F15, 9, A230097K15Rik, GPAT-3, mGPAT3 |
Location: |
Chr.5:100845710-100899102 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agpat9 | NM_172715.3 | NP_766303.1 | ||
| AK030171.1 | BAC26820.1 | |||
| AK138410.1 | BAE23647.1 | |||
| AK160261.1 | BAE35719.1 | |||
| BC096769.1 | AAH96769.1 | |||
| BC138228.1 | ||||
| BC145669.1 | ||||
| CN719213.1 |
More Information
Set Membership: |
|

Genomic Map