Assay Details
Target Gene Details
Entrez Gene ID: | 231872 |
Gene Name: | aminoacyl tRNA synthetase complex-interacting multifunctional protein 2 |
Gene Aliases: |
AA407136, AA407225, Aimp2(p38), Jtv1 |
Location: |
Chr.5:143902704-143909839 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aimp2 | NM_001172146.1 | 1 | 159 | NP_001165617.1 |
| NM_146165.2 | NP_666277.1 | |||
| BC024410.1 | 1 | 132 | AAH24410.1 | |
| BC024480.1 | AAH24480.1 | |||
| BC026958.1 | 1 | 98 | AAH26958.1 | |
| BC026972.1 | 1 | 112 | AAH26972.1 | |
| BY202241.1 | 1 | 160 | ||
| CF732613.1 | 1 | 81 | ||
| CJ051555.1 |
More Information
Set Membership: |
|

Genomic Map