Assay Details
Target Gene Details
Entrez Gene ID: | 12959 |
Gene Name: | crystallin, beta A4 |
Gene Aliases: |
- |
Location: |
Chr.5:112246492-112263555 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryba4 |
Target Gene Details
Entrez Gene ID: | 12960 |
Gene Name: | crystallin, beta B1 |
Gene Aliases: |
3110006K12Rik, BB1CRY |
Location: |
Chr.5:112254477-112269582 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crybb1 | NM_023695.2 | NP_076184.1 | ||
AF106853.1 | AAD42048.1 | |||
AK014012.1 | BAB29113.1 | |||
AK053869.1 | BAC35565.1 | |||
AK076443.1 | ||||
BC029012.1 | AAH29012.1 |
