Assay Details
Target Gene Details
Entrez Gene ID: | 171209 |
Gene Name: | acid-sensing (proton-gated) ion channel 3 |
Gene Aliases: |
AW742291, Accn3, DRASIC, SLNAC1, TNAC1 |
Location: |
Chr.5:24410289-24417834 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asic3 | NM_183000.2 | NP_892045.2 | ||
| AK028403.1 | ||||
| AY261387.1 | AAP88539.1 |
Target Gene Details
Entrez Gene ID: | 12568 |
Gene Name: | cyclin-dependent kinase 5 |
Gene Aliases: |
AW048668, Crk6 |
Location: |
Chr.5:24407597-24423591 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5 |
More Information
Set Membership: |
|

Genomic Map