Assay Details
Target Gene Details
Entrez Gene ID: | 100986 |
Gene Name: | A kinase (PRKA) anchor protein (yotiao) 9 |
Gene Aliases: |
5730481H23Rik, AKAP-9, AKAP450, AW545847, C79026, G1-448-15, PRKA9, mKIAA0803, mei2-5, repro12 |
Location: |
Chr.5:3927417-4080204 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Akap9 | NM_194462.2 | NP_919444.2 | ||
AJ582913.1 | CAE46960.1 | |||
AK008727.1 | BAB25859.1 | |||
AK017707.1 | BAB30886.3 | |||
AK031740.1 | BAC27533.1 | |||
AK080442.1 | BAC37918.1 | |||
AK168404.1 | BAE40324.1 |
