Assay Details
Target Gene Details
Entrez Gene ID: | 319974 |
Gene Name: | autism susceptibility candidate 2 |
Gene Aliases: |
2700063G02Rik, A730011F23Rik, D830032G16Rik |
Location: |
Chr.5:131437682-132542343 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Auts2 | NM_177047.3 | 18 | 2899 | NP_796021.2 |
AK012475.1 | 2 | 112 | BAB28267.1 | |
AK135848.1 | 3 | 232 | BAE22692.1 | |
AK164158.1 | 14 | 2339 | ||
BC021509.1 | 11 | 974 | AAH21509.1 | |
BC058110.1 | 6 | 475 | AAH58110.1 |
