Assay Details
Target Gene Details
Entrez Gene ID: | 74610 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 8 |
Gene Aliases: |
4833412N02Rik, AA409895 |
Location: |
Chr.5:24393681-24409947 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb8 | NM_029020.2 | 15 | 2270 | NP_083296.2 |
| AF213391.1 | AAF31430.1 | |||
| AK014319.1 | 15 | 2001 | BAB29270.1 | |
| AK030624.1 | 15 | 1997 | BAC27052.1 | |
| AK038760.1 | 15 | 1998 | ||
| AK049152.1 | 15 | 2014 | BAC33571.1 | |
| AK076315.1 | 15 | 2021 | BAC36297.1 | |
| AK166704.1 | 15 | 2004 | BAE38958.1 | |
| AK170334.1 | ||||
| AK178570.1 | ||||
| AK184256.1 | ||||
| BC015301.1 | 15 | 2270 | AAH15301.1 |
Target Gene Details
Entrez Gene ID: | 12568 |
Gene Name: | cyclin-dependent kinase 5 |
Gene Aliases: |
AW048668, Crk6 |
Location: |
Chr.5:24407597-24423591 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5 |
More Information
Set Membership: |
|

Genomic Map