Assay Details
Target Gene Details
Entrez Gene ID: | 231842 |
Gene Name: | archaelysin family metallopeptidase 1 |
Gene Aliases: |
5330426I05, 6530401C20Rik |
Location: |
Chr.5:140724127-140753312 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amz1 | NM_173405.2 | NP_775581.1 | ||
| AJ879912.1 | CAI53757.1 | |||
| AK078332.1 | BAC37222.1 | |||
| AK170176.1 | BAE41617.1 | |||
| BC046428.1 | AAH46428.1 | |||
| BC129841.1 | ||||
| BC129842.1 |

Genomic Map