Assay Details
Target Gene Details
Entrez Gene ID: | 22385 |
Gene Name: | bromodomain adjacent to zinc finger domain, 1B |
Gene Aliases: |
C87820, WSTF, Wbscr9 |
Location: |
Chr.5:135187323-135246129 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz1b | NM_011714.2 | 14 | 3833 | NP_035844.2 |
AF084480.1 | 14 | 3792 | AAD08676.1 | |
AK037737.1 | 8 | 1041 | BAC29862.1 | |
BC141399.1 | 14 | 3736 |
