Assay Details
Target Gene Details
Entrez Gene ID: | 22385 |
Gene Name: | bromodomain adjacent to zinc finger domain, 1B |
Gene Aliases: |
C87820, WSTF, Wbscr9 |
Location: |
Chr.5:135187323-135246129 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Baz1b | NM_011714.2 | 8 | 2980 | NP_035844.2 |
AF084480.1 | 8 | 2939 | AAD08676.1 | |
AK037737.1 | 2 | 188 | BAC29862.1 | |
BC141399.1 | 8 | 2883 |
