Assay Details
Target Gene Details
Entrez Gene ID: | 27405 |
Gene Name: | ATP-binding cassette, sub-family G (WHITE), member 3 |
Gene Aliases: |
Abcp2, Mxr2 |
Location: |
Chr.5:104935057-104982802 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcg3 | NM_030239.2 | NP_084515.2 | ||
| AF324242.1 | AAK14241.1 | |||
| AK030831.1 | ||||
| AK051880.1 | BAC34799.1 | |||
| AK156626.1 | ||||
| BC138772.1 | ||||
| BC138773.1 |
More Information
Set Membership: |
|

Genomic Map