Assay Details
Target Gene Details
Entrez Gene ID: | 12568 |
Gene Name: | cyclin-dependent kinase 5 |
Gene Aliases: |
AW048668, Crk6 |
Location: |
Chr.5:24407597-24423591 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5 | NM_007668.3 | NP_031694.1 | ||
| AK051790.1 | BAC34769.1 | |||
| BC052007.1 | AAH52007.1 | |||
| D29678.1 | BAA06148.1 | |||
| X64604.1 | CAA45888.1 |
More Information
Set Membership: |
|

Genomic Map