Assay Details
Target Gene Details
Entrez Gene ID: | 54188 |
Gene Name: | cleavage and polyadenylation specific factor 4 |
Gene Aliases: |
30kDa, C79664 |
Location: |
Chr.5:145167213-145182041 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpsf4 | NM_001291248.1 | NP_001278177.1 | ||
| NM_001291249.1 | NP_001278178.1 | |||
| NM_178576.3 | NP_848671.1 | |||
| AF033201.1 | AAC53567.1 | |||
| AK046064.1 | BAC32587.1 | |||
| BC057067.1 | AAH57067.1 | |||
| CN723345.1 | ||||
| EU588393.1 | ||||
| EU588394.1 |
More Information
Set Membership: |
|

Genomic Map