Assay Details
Target Gene Details
Entrez Gene ID: | 320454 |
Gene Name: | transmembrane protease, serine 11g |
Gene Aliases: |
9930032O22Rik, Desc4 |
Location: |
Chr.5:86485877-86518600 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss11g | NM_177162.4 | 9 | 1125 | NP_796136.2 |
| AK036981.1 | 9 | 1126 | BAC29657.1 | |
| AK037029.1 | 5 | 623 | BAC29676.1 | |
| BC118611.1 | 9 | 1103 | ||
| BC118660.1 | 9 | 1102 |
More Information
Set Membership: |
|

Genomic Map