Assay Details
Target Gene Details
Entrez Gene ID: | 213990 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 3 |
Gene Aliases: |
AGAP-3, AV028425, Centg3, Crag, Crag., MRIP-1, cnt-g3 |
Location: |
Chr.5:24452177-24502047 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agap3 | NM_001256431.1 | NP_001243360.1 | ||
| NM_139153.2 | NP_631892.2 | |||
| AB078345.1 | BAC55242.1 | |||
| AF459091.1 | AAL68640.1 | |||
| CJ052480.1 | ||||
| CJ053222.1 |

Genomic Map