Assay Details
Target Gene Details
Entrez Gene ID: | 57914 |
Gene Name: | cytokine receptor-like factor 2 |
Gene Aliases: |
CRLM2, Ly114, Tpte2, Tslpr |
Location: |
Chr.5:109554709-109559017 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Crlf2 | NM_001164735.1 | NP_001158207.1 | ||
| NM_016715.4 | NP_057924.3 | |||
| AB031333.1 | BAA92159.1 | |||
| AB039945.1 | BAA92684.1 | |||
| AF201963.1 | AAF82189.1 | |||
| AF232936.1 | AAF81676.1 | |||
| AK010291.1 | BAB26827.2 | |||
| AK149928.1 | ||||
| AK170466.1 | BAE41814.1 | |||
| BC016580.1 | ||||
| BC023788.1 | AAH23788.1 |
More Information
Set Membership: |
|

Genomic Map