Assay Details
Target Gene Details
Entrez Gene ID: | 11867 |
Gene Name: | actin related protein 2/3 complex, subunit 1B |
Gene Aliases: |
41kDa, AA408064, AA408534, AA571392, AF007010, AW208418, L72, SOP2Hs, p41-ARC |
Location: |
Chr.5:145114256-145128186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arpc1b | NM_023142.2 | NP_075631.2 | ||
| AF162768.1 | AAD43352.1 | |||
| AK002274.1 | BAB21980.1 | |||
| AK005378.1 | BAB23985.1 | |||
| AK147731.1 | BAE28101.1 | |||
| AK148145.1 | BAE28373.1 | |||
| AK151911.1 | ||||
| AK153510.1 | ||||
| AK157098.1 | BAE33961.1 | |||
| AK159371.1 | BAE35028.1 | |||
| AK159477.1 | ||||
| AK167978.1 | BAE39970.1 | |||
| AK168774.1 | BAE40610.1 | |||
| AK170759.1 | BAE42010.1 | |||
| AK171364.1 | BAE42412.1 | |||
| AK172124.1 | BAE42835.1 | |||
| BC003441.1 | AAH03441.1 | |||
| BC010275.1 | AAH10275.1 | |||
| BC092051.1 | AAH92051.1 |
More Information
Set Membership: |
|

Genomic Map