Assay Details
Target Gene Details
Entrez Gene ID: | 12617 |
Gene Name: | centromere protein C1 |
Gene Aliases: |
CENP-C, Cenpc |
Location: |
Chr.5:86012018-86066019 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cenpc1 | NM_007683.3 | NP_031709.2 | ||
| BC150700.1 | ||||
| U03113.1 | AAC04314.1 |
More Information
Set Membership: |
|

Genomic Map