Assay Details
Target Gene Details
Entrez Gene ID: | 209354 |
Gene Name: | eukaryotic translation initiation factor 2B, subunit 1 (alpha) |
Gene Aliases: |
26kDa, D5Ertd406e, EIF2B, EIF2BA |
Location: |
Chr.5:124570214-124579057 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2b1 | NM_145371.3 | 8 | 731 | NP_663346.1 |
| AK088066.1 | 8 | 730 | ||
| AK133695.1 | 8 | 732 | BAE21787.1 | |
| AK198878.1 | 3 | 172 | ||
| BC003426.1 | 8 | 731 | AAH03426.1 |
More Information
Set Membership: |
|

Genomic Map