Assay Details
Target Gene Details
Entrez Gene ID: | 70693 |
Gene Name: | adhesion G protein-coupled receptor A2 |
Gene Aliases: |
3830613O22Rik, AU044632, Gpr125, Tem5-like |
Location: |
Chr.5:49959951-50058996 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgra2 | NM_133911.1 | 8 | 1201 | NP_598672.1 |
| AK014455.1 | 8 | 1173 | BAC25436.1 | |
| AK028929.1 | 8 | 1140 | ||
| BC052391.1 | 8 | 1201 | AAH52391.1 |
More Information
Set Membership: |
|

Genomic Map