Assay Details
Target Gene Details
Entrez Gene ID: | 170935 |
Gene Name: | glutamate receptor, ionotropic, delta 2 (Grid2) interacting protein 1 |
Gene Aliases: |
- |
Location: |
Chr.5:143357338-143391798 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grid2ip | NM_001159321.1 | NP_001152793.1 | ||
| NM_133355.1 | NP_579933.1 | |||
| AF099933.2 | AAG31020.1 | |||
| DQ193535.1 | ABB04525.1 |

Genomic Map