Assay Details
Target Gene Details
Entrez Gene ID: | 66383 |
Gene Name: | IscU iron-sulfur cluster scaffold homolog (E. coli) |
Gene Aliases: |
2310020H20Rik, AA407971, Nifu, Nifun |
Location: |
Chr.5:113772812-113778282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iscu | NM_025526.4 | NP_079802.1 | ||
| AK009021.1 | BAB26031.1 | |||
| AK009421.1 | ||||
| AK039834.1 | BAC30464.1 | |||
| AK083247.1 | BAC38830.1 | |||
| AK157864.1 | ||||
| BC028800.1 | AAH28800.1 | |||
| BC048409.1 | AAH48409.1 | |||
| BC096049.1 | AAH96049.1 |
More Information
Set Membership: |
|

Genomic Map