Assay Details
Target Gene Details
Entrez Gene ID: | 15234 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
C230052L06Rik, HGF/SF, NK1, NK2, SF, SF/HGF |
Location: |
Chr.5:16553335-16622408 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgf | NM_001289458.1 | 7 | 891 | NP_001276387.1 |
| NM_001289459.1 | 7 | 880 | NP_001276388.1 | |
| NM_001289461.1 | 6 | 698 | NP_001276390.1 | |
| NM_010427.5 | 7 | 863 | NP_034557.3 | |
| AK042121.1 | 7 | 809 | BAC31175.1 | |
| BC119228.1 | 7 | 750 | ||
| D10212.1 | 6 | 686 | BAA01064.1 | |
| D10213.1 | 6 | 671 | BAA01065.1 | |
| S71816.1 | 6 | 686 | AAB31855.1 | |
| X72307.1 | 6 | 747 | CAA51054.1 | |
| X84046.1 | 6 | 726 | CAA58865.1 |
More Information
Set Membership: |
|

Genomic Map