Assay Details
Target Gene Details
Entrez Gene ID: | 15891 |
Gene Name: | integrin binding sialoprotein |
Gene Aliases: |
BSP, BSP II, BSPII, Bsp2 |
Location: |
Chr.5:104299287-104311472 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ibsp | NM_008318.3 | NP_032344.2 | ||
| AK132371.1 | BAE21132.1 | |||
| AK162640.1 | BAE37003.1 | |||
| BB618282.1 | ||||
| BC045143.1 | AAH45143.1 | |||
| L20232.1 | AAA21726.1 | |||
| L23801.1 | AAA37326.1 |
More Information
Set Membership: |
|

Genomic Map