Assay Details
Target Gene Details
Entrez Gene ID: | 13483 |
Gene Name: | dipeptidylpeptidase 6 |
Gene Aliases: |
B930011P16Rik, D5Buc3, D5Buc4, D5Buc5, DPP VI, Dpp-6, Gm1377, In(5)6H-p, Peplb, Rw |
Location: |
Chr.5:26817357-27727500 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpp6 | NM_001136060.2 | 20 | 2118 | NP_001129532.1 |
| NM_001198886.1 | 20 | 2202 | NP_001185815.1 | |
| NM_010075.2 | 20 | 2453 | NP_034205.1 | |
| NM_207282.3 | 20 | 2306 | NP_997165.2 | |
| AF092506.1 | 17 | 1466 | AAC97365.1 | |
| AF092507.1 | 20 | 2461 | AAC97366.1 | |
| AK031903.1 | 20 | 2079 | ||
| AK158964.1 | 20 | 2307 | BAE34745.1 | |
| BC048383.1 | 20 | 1969 | AAH48383.1 | |
| BC085154.1 | 20 | 2202 | AAH85154.1 |
More Information
Set Membership: |
|

Genomic Map