Assay Details
Target Gene Details
Entrez Gene ID: | 68303 |
Gene Name: | family with sequence similarity 114, member A1 |
Gene Aliases: |
1190001N04Rik, 9130005N14Rik, Noxp20 |
Location: |
Chr.5:64970075-65041901 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam114a1 | NM_026667.3 | NP_080943.3 | ||
| AK007950.1 | ||||
| AK014725.1 | ||||
| AK017336.1 | BAB30694.1 | |||
| AK020262.1 | BAB32046.1 | |||
| AK030360.1 | BAC26919.1 | |||
| AM162548.1 | CAJ44264.1 | |||
| BC027235.1 | AAH27235.1 | |||
| BC110359.1 | AAI10360.1 |
Target Gene Details
Entrez Gene ID: | 21899 |
Gene Name: | toll-like receptor 6 |
Gene Aliases: |
- |
Location: |
Chr.5:64952030-64971173 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tlr6 |

Genomic Map