Assay Details
Target Gene Details
Entrez Gene ID: | 84652 |
Gene Name: | family with sequence similarity 126, member A |
Gene Aliases: |
AB030242, Drctnnb1a |
Location: |
Chr.5:23915276-24030780 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam126a | NM_053090.2 | 8 | 925 | NP_444320.2 |
| AK036680.1 | 1 | 1829 | ||
| AK155209.1 | 8 | 926 | BAE33120.1 | |
| AK161265.1 | 8 | 1119 | BAE36278.1 | |
| BC056381.1 | 8 | 864 | AAH56381.1 | |
| BC060692.1 | 8 | 912 | AAH60692.1 |

Genomic Map