Assay Details
Target Gene Details
Entrez Gene ID: | 57080 |
Gene Name: | general transcription factor II I repeat domain-containing 1 |
Gene Aliases: |
1700012P16Rik, BEN, Cream1, ESTM9, GTF3, Gtf2il, MusTRD1, Tg(Alb1-Myc)166.8Sst, WBSCR11, X83320 |
Location: |
Chr.5:134357661-134456716 on Build GRCm38 |
Assay Gene Location: | Within Intron 29 |
More Information
Set Membership: |
|

Genomic Map