Assay Details
Target Gene Details
Entrez Gene ID: | 107999 |
Gene Name: | GTP binding protein 6 (putative) |
Gene Aliases: |
AV119224, Pgbpll, pgpl |
Location: |
Chr.5:110103976-110108197 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtpbp6 | NM_145147.5 | NP_660129.2 | ||
AJ293619.1 | CAC82171.1 | |||
AK082192.1 | 1 | 228 | BAC38433.1 | |
AK144812.1 | BAE26076.1 | |||
AK152225.1 | BAE31052.1 | |||
AK152972.1 | BAE31629.1 | |||
AK159462.1 | BAE35103.1 |
