Assay Details
Target Gene Details
Entrez Gene ID: | 107999 |
Gene Name: | GTP binding protein 6 (putative) |
Gene Aliases: |
AV119224, Pgbpll, pgpl |
Location: |
Chr.5:110103976-110108197 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp6 | NM_145147.5 | NP_660129.2 | ||
| AJ293619.1 | CAC82171.1 | |||
| AK082192.1 | BAC38433.1 | |||
| AK144812.1 | BAE26076.1 | |||
| AK152225.1 | BAE31052.1 | |||
| AK152972.1 | BAE31629.1 | |||
| AK159462.1 | BAE35103.1 | |||
| BC027821.1 | AAH27821.1 |
Target Gene Details
Entrez Gene ID: | 403178 |
Gene Name: | phosphatidylinositol-specific phospholipase C, X domain containing 1 |
Gene Aliases: |
A330045H12Rik, Gm576 |
Location: |
Chr.5:110099225-110105953 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcxd1 | NM_001281812.1 | 7 | 2436 | NP_001268741.1 |
| NM_207279.2 | 8 | 2601 | NP_997162.2 | |
| AK149435.1 | 7 | 2437 |

Genomic Map