Assay Details
Target Gene Details
Entrez Gene ID: | 23980 |
Gene Name: | phosphatidylethanolamine binding protein 1 |
Gene Aliases: |
HCNP, Pbp, Pbp1, Pbpr, Rkip |
Location: |
Chr.5:117282651-117287564 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pebp1 | NM_018858.2 | NP_061346.2 | ||
| AB046417.1 | BAB03276.1 | |||
| AF300422.1 | AAG25635.1 | |||
| AK088212.1 | BAC40214.1 | |||
| AK143897.1 | BAE25588.1 | |||
| AK149815.1 | BAE29101.1 | |||
| AK159842.1 | BAE35420.1 | |||
| AK168790.1 | BAE40623.1 | |||
| AK182328.1 | ||||
| BC008169.1 | AAH08169.1 | |||
| BC083063.1 | AAH83063.1 | |||
| BC089332.1 | AAH89332.1 | |||
| U43206.1 | AAB06983.1 |
More Information
Set Membership: |
|

Genomic Map