Assay Details
Target Gene Details
Entrez Gene ID: | 19079 |
Gene Name: | protein kinase, AMP-activated, beta 1 non-catalytic subunit |
Gene Aliases: |
1300015D22Rik, AU021155, E430008F22 |
Location: |
Chr.5:116013586-116024503 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkab1 | NM_031869.2 | NP_114075.1 | ||
| AK088250.1 | BAC40237.1 | |||
| AK153552.1 | BAE32089.1 | |||
| AK180257.1 | ||||
| BC016398.1 | AAH16398.1 |
More Information
Set Membership: |
|

Genomic Map