Assay Details
Target Gene Details
Entrez Gene ID: | 66627 |
Gene Name: | 2-oxoglutarate and iron-dependent oxygenase domain containing 2 |
Gene Aliases: |
1300006G11Rik, 5730405M13Rik, AU041636, AW552395 |
Location: |
Chr.5:124112058-124115480 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogfod2 | NM_025671.2 | NP_079947.1 | ||
| AK004916.1 | 2 | 229 | ||
| AK004943.1 | BAB23687.1 | |||
| AK017497.1 | BAC25517.1 | |||
| AK018534.1 | BAB31259.1 | |||
| AK168946.1 | ||||
| BC022762.1 | AAH22762.1 | |||
| BC069872.1 | AAH69872.1 |
More Information
Set Membership: |
|

Genomic Map