Assay Details
Target Gene Details
Entrez Gene ID: | 18778 |
Gene Name: | phospholipase A2, group IB, pancreas |
Gene Aliases: |
Pla2a, sPLA2IB |
Location: |
Chr.5:115466241-115474717 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g1b | NM_011107.1 | NP_035237.1 | ||
| AF094610.1 | AAG27064.1 | |||
| AF097637.1 | AAD19896.1 | |||
| AF162712.1 | AAD45806.1 | |||
| AF187852.1 | AAF44297.1 | |||
| AK007730.1 | BAB25218.1 | |||
| AK007797.1 | BAB25263.1 | |||
| AK008331.1 | BAB25608.1 | |||
| AK008664.1 | BAB25819.1 | |||
| AK008668.1 | BAB25822.1 | |||
| AK008841.1 | BAB25922.1 | |||
| AK008934.1 | BAB25976.1 | |||
| AK008936.1 | BAB25978.1 | |||
| AK009314.1 | BAB26212.1 | |||
| AK019009.1 | ||||
| AK028104.1 | BAC25749.1 | |||
| AK028134.1 | BAC25763.1 | |||
| BC145908.1 | ||||
| BC145910.1 |
More Information
Set Membership: |
|

Genomic Map