Assay Details
Target Gene Details
Entrez Gene ID: | 17220 |
Gene Name: | minichromosome maintenance deficient 7 (S. cerevisiae) |
Gene Aliases: |
AI747533, Mcmd7, mCDC47 |
Location: |
Chr.5:138164589-138171862 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm7 | NM_008568.2 | NP_032594.1 | ||
| AK133091.1 | BAE21506.1 | |||
| AK144980.1 | BAE26164.1 | |||
| AK149913.1 | ||||
| AK150059.1 | BAE29273.1 | |||
| AK150066.1 | ||||
| AK154052.1 | BAE32342.1 | |||
| BC065164.1 | AAH65164.1 | |||
| D26091.1 | BAA05084.1 |
More Information
Set Membership: |
|

Genomic Map