Assay Details
Target Gene Details
Entrez Gene ID: | 18647 |
Gene Name: | cyclin-dependent kinase 14 |
Gene Aliases: |
Pftk1, mKIAA0834 |
Location: |
Chr.5:4803384-5380251 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk14 | NM_011074.2 | NP_035204.2 | ||
| AF033655.1 | AAB87504.1 | |||
| AK129226.1 | BAC98036.1 | |||
| AK138437.1 | BAE23662.1 | |||
| AK139386.1 | BAE23987.1 | |||
| AK141535.1 | BAE24722.1 | |||
| AK165149.1 | BAE38049.1 | |||
| BC068134.1 | AAH68134.1 | |||
| U62391.1 | AAB70455.1 |
More Information
Set Membership: |
|

Genomic Map