Assay Details
Target Gene Details
Entrez Gene ID: | 18647 |
Gene Name: | cyclin-dependent kinase 14 |
Gene Aliases: |
Pftk1, mKIAA0834 |
Location: |
Chr.5:4803384-5380251 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk14 | NM_011074.2 | 3 | 460 | NP_035204.2 |
| AF033655.1 | 3 | 419 | AAB87504.1 | |
| AK016370.1 | 3 | 402 | BAB30208.1 | |
| AK045083.1 | 3 | 461 | BAC32212.1 | |
| AK051283.1 | 3 | 434 | BAE43346.1 | |
| AK051411.1 | 3 | 403 | BAC34630.1 | |
| AK083269.1 | 2 | 615 | BAC38836.1 | |
| AK083790.1 | 3 | 403 | BAC39020.1 | |
| AK087398.1 | 2 | 231 | BAC39861.1 | |
| AK129226.1 | 3 | 287 | BAC98036.1 | |
| AK138437.1 | 2 | 701 | BAE23662.1 | |
| AK139386.1 | 2 | 621 | BAE23987.1 | |
| AK141535.1 | 2 | 389 | BAE24722.1 | |
| AK165149.1 | 2 | 305 | BAE38049.1 | |
| BC068134.1 | 2 | 664 | AAH68134.1 | |
| U62391.1 | 2 | 314 | AAB70455.1 |
More Information
Set Membership: |
|

Genomic Map