Assay Details
Target Gene Details
Entrez Gene ID: | 19181 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, ATPase 2 |
Gene Aliases: |
- |
Location: |
Chr.5:21785283-21803784 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmc2 | NM_011188.3 | NP_035318.1 | ||
| AK005083.1 | BAB23807.1 | |||
| AK076874.1 | BAC36516.1 | |||
| AK146919.1 | BAE27531.1 | |||
| AK153413.1 | BAE31973.1 | |||
| AK168548.1 | BAE40423.1 | |||
| AK216249.1 | ||||
| BC005462.1 | AAH05462.1 | |||
| U61283.1 | AAB51069.1 |
More Information
Set Membership: |
|

Genomic Map