Assay Details
Target Gene Details
Entrez Gene ID: | 269642 |
Gene Name: | N-acetyltransferase 8-like |
Gene Aliases: |
1110038O08Rik, Shati |
Location: |
Chr.5:33995984-34005916 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat8l | NM_001001985.3 | NP_001001985.3 | ||
| AK147307.1 | BAE27836.1 | |||
| AK147594.1 | BAE28014.1 | |||
| AK147703.1 | BAE28084.1 | |||
| BC075616.1 | AAH75616.1 | |||
| DQ174094.1 |
More Information
Set Membership: |
|

Genomic Map