Assay Details
Target Gene Details
Entrez Gene ID: | 20022 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide J |
Gene Aliases: |
14.5kDa, Polr2i, Rpb11a, Rpo2-4 |
Location: |
Chr.5:136116691-136122947 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2j | NM_011293.2 | NP_035423.2 | ||
| AK158610.1 | BAE34581.1 | |||
| AK158644.1 | BAE34594.1 |
More Information
Set Membership: |
|

Genomic Map