Assay Details
Target Gene Details
Entrez Gene ID: | 100609 |
Gene Name: | NOL1/NOP2/Sun domain family, member 5 |
Gene Aliases: |
9830109N13Rik, AI326939, Nol1r, Wbscr20, Wbscr20a |
Location: |
Chr.5:135369930-135376805 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nsun5 | NM_145414.2 | 9 | 1253 | NP_663389.2 |
AF412029.1 | 9 | 1246 | AAM62311.1 | |
AK079400.1 | 9 | 1254 | BAC37634.1 | |
AK149521.1 | 9 | 1235 | BAE28935.1 | |
AK156651.1 | 9 | 1248 | BAE33793.1 | |
BC051209.1 | 5 | 685 | AAH51209.1 |
