Assay Details
Target Gene Details
Entrez Gene ID: | 20005 |
Gene Name: | ribosomal protein L9 |
Gene Aliases: |
- |
Location: |
Chr.5:65388364-65391431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl9 | NM_011292.2 | NP_035422.1 | ||
| AF260271.1 | AAF70508.1 | |||
| AK012331.1 | BAB28167.1 | |||
| AK012445.1 | BAB28244.1 | |||
| AK017388.1 | BAB30725.1 | |||
| AK017427.1 | BAB30739.1 | |||
| AK050296.1 | ||||
| AK053431.1 | ||||
| AK084278.1 | BAC39154.1 | |||
| AK088166.1 | BAC40185.1 | |||
| AK153562.1 | BAE32097.1 | |||
| AK159980.1 | BAE35531.1 | |||
| AK168326.1 | BAE40265.1 | |||
| AK169217.1 | BAE40988.1 | |||
| BC013165.1 | AAH13165.1 | |||
| BC081435.1 | AAH81435.1 | |||
| BC083166.1 | AAH83166.1 | |||
| BC083329.1 | AAH83329.1 | |||
| BC086937.1 | AAH86937.1 | |||
| BC089319.1 | AAH89319.1 | |||
| BC093526.1 | AAH93526.1 | |||
| BY072215.1 |
More Information
Set Membership: |
|

Genomic Map