Assay Details
Target Gene Details
Entrez Gene ID: | 20005 |
Gene Name: | ribosomal protein L9 |
Gene Aliases: |
- |
Location: |
Chr.5:65388364-65391431 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpl9 | NM_011292.2 | NP_035422.1 | ||
AF260271.1 | AAF70508.1 | |||
AK012331.1 | BAB28167.1 | |||
AK012445.1 | BAB28244.1 | |||
AK017388.1 | BAB30725.1 | |||
AK017427.1 | BAB30739.1 | |||
AK050296.1 | 2 | 308 | ||
AK053431.1 | ||||
AK084278.1 | BAC39154.1 | |||
AK088166.1 | BAC40185.1 | |||
AK153562.1 | BAE32097.1 | |||
AK159980.1 | BAE35531.1 | |||
AK168326.1 | BAE40265.1 | |||
AK169217.1 | BAE40988.1 | |||
BC013165.1 | AAH13165.1 | |||
BC081435.1 | AAH81435.1 | |||
BC083166.1 | AAH83166.1 | |||
BC083329.1 | AAH83329.1 | |||
BC086937.1 | AAH86937.1 | |||
BC089319.1 | AAH89319.1 | |||
BC093526.1 | AAH93526.1 | |||
BY072215.1 | ||||
U17331.1 | AAA85685.1 | |||
U17332.1 | AAA85686.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
